Anti-AK7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513836
CAPIPA5-13836EA 2561.45 CAD
CAPIPA5-13836
Anti-AK7 Rabbit Polyclonal Antibody
This antibody is predicted to react with non-human primate based on sequence homology. The adenylate kinases (AK) are a family of structurally and functionally related enzymes that catalyze a similar reaction, MgNTP + AMP = MgNDP + ADP (N = A or G). The AK enzymes are important for maintenance of homeostasis of the adenine and guanine nucleotide pools. AK1 is a cytosolic enzyme for which ATP is the substrate. AK2 catalyzes the same reaction as AK1, but it is localized in the mitochondrial intermembrane space. AK3 is present in the mitochondrial matrix and prefers GTP over ATP as the substrate.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: AK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR