Anti-DCK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513855
CAPIPA5-13855EA 2561.45 CAD
CAPIPA5-13855
Anti-DCK Rabbit Polyclonal Antibody
Deoxycytidine kinase is responsible for the phosphorylation of several deoxyribonucleosides and their analogs. Deficiency of this enzyme activity is associated with resistance to antiviral and anticancer chemotherapeutic agents, whereas increased enzyme activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. It is the rate limiting enzyme in the activation of many important anticancer and retroviral drugs and its activity is often decreased in cells that are resistant to cytosine arabinoside.

Immunocytochemistry: 1: 10 - 1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Immunoprecipitation: 1:500-1:1000, Western Blot: 1:1000

Type: Primary
Antigen: DCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR