Anti-PGK1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513863
CAPIPA5-13863EA 2573.22 CAD
CAPIPA5-13863
Anti-PGK1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with non-human primate and rat based on sequence homology. Also known as ATP:3-phosphoglycerate 1-phosphotransferase (EC 2.7.2.3), this major enzyme in glycolysis catalyzes the reversible conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate, generating one molecule of ATP. New blood vessel formation or angiogenesis is critical for tumor expansion and metastasis. Lay et al. (2000) showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PGK1 (phosphoglycerate kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR