Anti-ACVR1C Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513883
CAPIPA5-13883EA 2561.45 CAD
CAPIPA5-13883
Anti-ACVR1C Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. ACVR1C, a serine/threonine protein kinase, is a type I receptor for the TGFB family of signaling molecules. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. ACVR1C is a receptor for activin AB, activin B and NODAL. This protein plays a role in cell differentiation, growth arrest and apoptosis.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: ACVR1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR