Anti-ADCK5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513893
CAPIPA5-13893EA 2561.45 CAD
CAPIPA5-13893
Anti-ADCK5 Rabbit Polyclonal Antibody
ADCK5 is member of the AarF domain containing kinase family.

Western Blot: 1:1000

Type: Primary
Antigen: ADCK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR