Anti-AMHR2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513901
CAPIPA5-13901EA 2444.36 CAD
CAPIPA5-13901
Anti-AMHR2 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. The AMH receptor (AMHR or AMHR2) is a serine/threonine kinase with a single transmembrane domain belonging to the family of type II receptors for TGF-beta-related proteins. Anti-Mullerian hormone (AMH) and its receptor are involved in the regression of Mullerian ducts in male fetuses. Male sex differentiation is mediated by 2 discrete hormones produced by the fetal testis. Testosterone, produced by Leydig cells, virilizes the external genitalia and promotes prostatic growth; anti-Mullerian hormone (AMH) results in regression of Mullerian ducts which would otherwise differentiate into the uterus and fallopian tubes.

Western Blot: 1:1000

Type: Primary
Antigen: AMHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR