Anti-CABC1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513906
CAPIPA5-13906EA 2231.48 CAD
CAPIPA5-13906
Anti-CABC1 Rabbit Polyclonal Antibody
CABC1 is one of several proteins for which expression is induced by p53 expression. The S. pombe homolog, ABC1, is a chaperone-like protein essential for the proper conformation and functioning of protein complexes in the respiratory chain.

Western Blot: 1:1000

Type: Primary
Antigen: CABC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR