Anti-CDK20 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513913
CAPIPA5-13913EA 2814.74 CAD
CAPIPA5-13913
Anti-CDK20 Rabbit Polyclonal Antibody
CCRK contains a kinase domain most closely related to the cyclin-dependent protein kinases. This protein is involved in cell growth and activates CDK2, a kinase involved in the control of the cell cycle, by phosphorylating residue Thr-160.

Western Blot: 1:1000

Type: Primary
Antigen: CDK20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR