Anti-EIF2AK4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513924
CAPIPA5-13924EA 2573.22 CAD
CAPIPA5-13924
Anti-EIF2AK4 Rabbit Polyclonal Antibody
Antibodies
GCN2 (EIF2AK4) belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 to downregulate protein synthesis in response to varied cellular stresses.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: EIF2AK4 (eukaryotic translation initiation factor 2 alpha kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR