Anti-Guanylyl Cyclase alpha 1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513928
CAPIPA5-13928EA 2573.22 CAD
CAPIPA5-13928
Anti-Guanylyl Cyclase alpha 1 Rabbit Polyclonal Antibody
Antibodies
Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: Guanylyl Cyclase alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR