Anti-MKNK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513953
CAPIPA5-13953EA 2801.79 CAD
CAPIPA5-13953
Anti-MKNK2 Rabbit Polyclonal Antibody
Antibodies
MKNK2 may play a role in the response to environmental stress and cytokines. This protein appears to regulate transcription by phosphorylating EIF4E, thus increasing the affinity of this protein for the 7-methylguanosine-containing mRNA cap.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MKNK2 (MAP kinase interacting serine/threonine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR