Anti-PANK3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513963
CAPIPA5-13963EA 2561.45 CAD
CAPIPA5-13963
Anti-PANK3 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. PANK3 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PANK3 (pantothenate kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR