Anti-PBK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513968
CAPIPA5-13968EA 2561.45 CAD
CAPIPA5-13968
Anti-PBK Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. PBK is a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase (MAPKK) family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: PBK (PDZ binding kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR