Anti-PGK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513975
CAPIPA5-13975EA 2561.45 CAD
CAPIPA5-13975
Anti-PGK2 Rabbit Polyclonal Antibody
This antibody is predicted to react with non-human primate based on sequence homology. PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PGK2 (phosphoglycerate kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR