Anti-PIM3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513976
CAPIPA5-13976EA 2801.79 CAD
CAPIPA5-13976
Anti-PIM3 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. PIM3 is a memberof the Ser/Thr protein kinase family that may be involved in cell cycle progression and anti-apoptosis process. This protein has been implicated in proliferation of human hepatoma cell lines. PIM3 is widely expressed, with the exception of no expression observed in colon, thymus, and small intestine. PIM3 is expressed in human hepatoma cell lines but not in normal liver tissues.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: PIM3 (pim-3 oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR