Anti-ELF4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513998
CAPIPA5-13998EA 2573.22 CAD
CAPIPA5-13998
Anti-ELF4 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and mouse based on sequence homology. ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ELF4 (E74-like factor 4 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR