Anti-UCK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA514010
CAPIPA5-14010EA 2882.37 CAD
CAPIPA5-14010
Anti-UCK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: UCK2 (uridine-cytidine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR