Anti-HMGB2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA514015
CAPIPA5-14015EA 2444.36 CAD
CAPIPA5-14015
Anti-HMGB2 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, porcine and rat based on sequence homology. HMGB2 is a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HMGB2 (high mobility group box 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR