Anti-PIAS2 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18489
CAPIPA5-18489EA 2943.07 CAD
CAPIPA5-18489
Anti-PIAS2 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.

Recommended Dilutions: Immunohistochemistry: 3-5µg/ml

Type: Primary
Antigen: PIAS2 (protein inhibitor of activated STAT, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR