Anti-B7-H4 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18771
CAPIPA5-18771EA 3034.47 CAD
CAPIPA5-18771
Anti-B7-H4 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. B7H4 belongs to the B7 family of costimulatory proteins. These proteins are expressed on the surface of antigen-presenting cells and interact with ligands on T lymphocytes.

Western Blot: 0.5-1.5µg/ml

Type: Primary
Antigen: B7-H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR