Anti-NOD1 Goat Polyclonal Antibody

Supplier: Pierce Biotechnology
PA5-18027
CAPIPA5-18027EA 2942.21 CAD
CAPIPA5-18027
Anti-NOD1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This gene encodes a member of the NOD family. This member is a cytosolic protein. It contains an N-terminal caspase recruitment domain in its C terminus. The CARD is involved in apoptotic signaling, LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid. Multiple alternatively spliced transcript variants differring in the 5' UTR have been described, but the full-length nature of these variants has not been determined.

Recommended Dilutions: Western Blot: 0.5-1.5 ug/ml

Type: Primary
Antigen: NOD1 (nucleotide-binding oligomerization domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR