Would you like to log in again?
Stainless Steel Framed Glass Hinged Door Wall Unit, Kloppenberg & Co.
Supplier: KLOPPENBERG & CO.
WE241330 WE181330
Stainless Steel Framed Glass Hinged Door Wall Unit comprised of T304 stainless steel with a #4 brushed satin finish
This unit has a solid back, a single adjustable shelf and can be modified for a right or left swing. Overall dimensions are 18"x13"x30". As with all Kloppenberg products, the stainless steel used is made of an industry standard minimum of 80% recyclable materials, ensuring this product is not only long lasting but eco-friendly.
Certifications: SEFA Section 8 tested and approved.
ORDER
Expand All / Collapse All
Depth Height Width Supplier No. VWR Catalog Number Unit Price Quantity
13" 30" 18" WE181330 10039-596 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
13" 30" 24" WE241330 10039-598 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally preferable attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty