Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flame Resistant CC1 Contractor Coveralls with Max Comfort™ Material, Red, MCR Safety

Supplier: MCR SAFETY
CC1R40 CC1R42 CC1R60 CC1R36 CC1R58 CC1R38 CC1R54 CC1R56 CC1R34 CC1R38T CC1R50 CC1R48T CC1R52 CC1R50T CC1R60T CC1R52T CC1R62T CC1R40T CC1R48 CC1R54T CC1R64T CC1R42T CC1R56T CC1R44 CC1R44T CC1R66T CC1R58T CC1R36T CC1R46T CC1R46
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics

  • 100% cotton material, lifetime FR guarantee
  • Two needle reinforced stitching on major seams for durability
  • Heavy-duty two-way YKK® brass break-away zipper with Neomex® tape
  • Multiple pockets on chest, hips, back and leg
  • NFPA 2112, NFPA 70E, UL Certified

The CC1 Coverall features a comfortable, 100% cotton material that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.

Certifications: NFPA 2112, NFPA 70E, UL Certified.



 

ORDER

Expand All / Collapse All
Description Size Material Length Thickness Color Supplier No. VWR Catalog Number Unit Price Quantity
Max Comfort™ CC1 Contractor Coverall 34 100% Cotton Regular 7 oz. Red CC1R34 10812-696 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 36 100% Cotton Regular 7 oz. Red CC1R36 10812-698 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 36T 100% Cotton Tall 7 oz. Red CC1R36T 10812-700 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 38 100% Cotton Regular 7 oz. Red CC1R38 10812-702 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 38T 100% Cotton Tall 7 oz. Red CC1R38T 10812-704 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 40 100% Cotton Regular 7 oz. Red CC1R40 10812-706 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 40T 100% Cotton Tall 7 oz. Red CC1R40T 10812-708 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 42 100% Cotton Regular 7 oz. Red CC1R42 10812-710 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 42T 100% Cotton Tall 7 oz. Red CC1R42T 10812-712 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 44 100% Cotton Regular 7 oz. Red CC1R44 10812-714 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 44T 100% Cotton Tall 7 oz. Red CC1R44T 10812-716 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 46 100% Cotton Regular 7 oz. Red CC1R46 10812-718 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 46T 100% Cotton Tall 7 oz. Red CC1R46T 10812-720 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 48 100% Cotton Regular 7 oz. Red CC1R48 10812-722 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 48T 100% Cotton Tall 7 oz. Red CC1R48T 10812-724 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 50 100% Cotton Regular 7 oz. Red CC1R50 10812-726 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 50T 100% Cotton Tall 7 oz. Red CC1R50T 10812-728 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 52 100% Cotton Regular 7 oz. Red CC1R52 10812-730 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 52T 100% Cotton Tall 7 oz. Red CC1R52T 10813-518 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 54 100% Cotton Regular 7 oz. Red CC1R54 10813-520 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 54T 100% Cotton Tall 7 oz. Red CC1R54T 10813-522 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 56 100% Cotton Regular 7 oz. Red CC1R56 10813-524 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 56T 100% Cotton Tall 7 oz. Red CC1R56T 10812-732 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 58 100% Cotton Regular 7 oz. Red CC1R58 10812-734 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 58T 100% Cotton Tall 7 oz. Red CC1R58T 10812-736 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 60 100% Cotton Regular 7 oz. Red CC1R60 10812-738 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 60T 100% Cotton Tall 7 oz. Red CC1R60T 10812-740 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 62T 100% Cotton Tall 7 oz. Red CC1R62T 10812-742 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 64T 100% Cotton Tall 7 oz. Red CC1R64T 10812-744 Each Retrieving
UL Listed
Max Comfort™ CC1 Contractor Coverall 66T 100% Cotton Tall 7 oz. Red CC1R66T 10812-746 Each Retrieving
UL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty