Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bottle Rocket Launcher

Supplier: EISCO SCIENTIFIC LLC
PH1120143 4301
The Bottle Rocket Launcher uses the power of compressed air and water for liftoff.

  • Launcher Can Fire Bottles over 100 ft. into the Air
  • Corded Release Mechanism for Remote Launches Included
  • Compressed Air and Water Function as Fuel
  • Pumps Pressurize Bottles

When attached to a sturdy base, the launcher can fire 2 L plastic bottles over 100 feet in the air.

Using water as liquid fuel, pressurize the bottle with a bicycle pump (not included). Remotely launch from a safe distance with the corded release mechanism.



 

ORDER

Expand All / Collapse All

Launcher
Description Supplier No. VWR Catalog Number Unit Price Quantity
Bottle Rocket Launcher PH1120143 470219-532 Each Retrieving

Bicycle Pump for Launcher
Description Height Supplier No. VWR Catalog Number Unit Price Quantity
Bicycle Pump with Ø 11/4" 171/4" 4301 470089-326 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping