Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCDC155 Rabbit Polyclonal Antibody

Supplier: Abgent

Western Blot: 1:1000

Type: Primary
Antigen: CCDC155
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
400 µL AP20386C 89513-760 Each (1mg) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Amino acid number 159 to 185
Antibody type Primary
Antigen name coiled-coil Domain Containing 155
Antigen symbol CCDC155
Antigen synonyms KASH5
Clonality Polyclonal
Concentration 0.5mg/ml
Conjugation Unconjugated
ELISA Yes
Epitope Center
Gene ID 147872
Host Rabbit
Immunogen This CCDC155 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 159-185 amino acids from the Central region of human CCDC155.
Molecular Weight 62.783 kDa
Purification Protein A & Peptide affinity purification
Reactivity Human
Sequence RRLVGENAKLQRC
Shipping Temperature 4°C
Storage Buffer PBS with 0.09% (W/V) sodium azide, pH7.4
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C in small aliquots to prevent freeze-thaw cycles
Western Blot Yes
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty