Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MFN2 Mouse Monoclonal Antibody [clone: N153/5]

Supplier: Biolegend
820301
Type: Primary
Antigen: MFN2
Clonality: Monoclonal
Clone: N153/5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 820301 CA10816-822 Each (100µl) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MFN2
Antigen name Mitofusin 2
Reactivity Mouse, Rat
Clonality Monoclonal
Antibody type Primary
Clone N153/5
Host Mouse
Isotype IgG2a
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms HSG, Transmembrane GTPase MFN2, KIAA0214, CMT2A, MARF, CPRP1, Mitofusin-2, CMT2A2, MFN2
Gene ID 170731
Concentration 1 mg/ml
Immunogen This monoclonal antibody was raised against a fusion protein corresponding to amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2.
Purification The antibody was purified by affinity chromatography.
Form Phosphate-buffered solution.
Storage temperature Store at -20°C.
Shipping temperature Dry Ice

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty