Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP9 Mouse Monoclonal Antibody [clone: L51/82]

Supplier: Biolegend
819701
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: L51/82
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 819701 CA10816-810 Each (100µl) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MMP9
Antigen name Matrix Metalloproteinase 9
Reactivity Mouse, Human, Rat
Clonality Monoclonal
Antibody type Primary
Clone L51/82
Host Mouse
Isotype IgG2a
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms 92kDa type IV collagenase), CLG4B, 92kDa gelatinase, macrophage gelatinase, GELB, type V collagenase, MMP-9, matrix metalloproteinase 9 (gelatinase B, 92 kDa gelatinase, MANDP2, 92 kDa type IV collagenase, matrix metalloproteinase-9
Gene ID 81687
Concentration 1 mg/ml
Immunogen This monoclonal antibody was raised against a full-length fusion protein corresponding to amino acids 1-708 of rat MMP9 protein.
Purification The antibody was purified by affinity chromatography.
Form Phosphate-buffered solution.
Storage temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty