Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDK7 Mouse Monoclonal Antibody [clone: K1]

Supplier: Biolegend
922402
Type: Primary
Antigen: CDK7
Clonality: Monoclonal
Clone: K1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mL 922402 CA10818-176 Each (1ml) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CDK7
Antigen name Cyclin-dependent Kinase 7
Reactivity Human, Monkey
Clonality Monoclonal
Antibody type Primary
Clone K1
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms CDK-activating kinase, 39 kDa protein kinase, STK1, MO15, CAK1, Cell division protein kinase 7, TFIIH basal transcription factor complex kinase subunit, CDK7, CAK, P39 Mo15
Gene ID 1022
Format Ascites
Immunogen This antibody was developed using spleen lymphocytes from mice fused with OVCAR-3, an ovarian tumor cell line derived from poorly differentiated adenocarcinoma of the ovary.
Purification Ascites
Form Ascites (in PBS with BSA + 0.1% NaN3).
Storage temperature Store between 2°C and 8°C.
Shipping temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty