Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABCC1 Rat Monoclonal Antibody [clone: MRPr1]

Supplier: Biolegend
907201
Type: Primary
Antigen: ABCC1
Clonality: Monoclonal
Clone: MRPr1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 µL 907201 CA10817-356 Each (500µl) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ABCC1
Antigen name ATP-binding Cassette, Subfamily C (CFTR/MRP), Member 1
Reactivity Human
Clonality Monoclonal
Antibody type Primary
Clone MRPr1
Host Rat
Isotype IgG2a
ImmunoChemistry Yes
Antigen synonyms multiple drug resistance protein 1, ATP-binding cassette, MRP1, DKFZp781G125, GS-X, sub-family C, LTC4 transporter, ABCC1, MRP, ABCC, multiple drug resistance-associated protein, ABC29, member 1, multidrug resistance protein, sub-family C (CFTR/MRP), DKFZp686N04233, leukotriene C(4) transporter
Gene ID 4363
Format Tissue Culture Supernatant
Immunogen This antibody was developed against a bacterial fusion protein of MRP containing a segment of 168 amino acids in the amino-proximal half of the protein.
Purification Tissue Culture Supernatant
Form Tissue Culture Supernatant (in PBS with BSA + 0.1% NaN3).
Storage temperature Store between 2°C and 8°C.
Shipping temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty