Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABCC3 Mouse Monoclonal Antibody [clone: M3II-21]

Supplier: Biolegend

Type: Primary
Antigen: ABCC3
Clonality: Monoclonal
Clone: M3II-21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 µL 907401 CA10817-360 Each (500µl) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antibody type Primary
Antigen name ATP-binding cassette, Subfamily C (CFTR/MRP), Member 3
Antigen symbol ABCC3
Antigen synonyms ABC31, MOAT-D, MRP3, EST90757, cMOAT2, MLP2
Clonality Monoclonal
Clone M3II-21
Concentration .25 mg/ml
Form Phosphate-buffered solution with BSA + 0.1% NaN3
Format Tissue Culture Supernatant
Gene ID 8714
Host Mouse
ImmunoChemistry Yes
Immunogen This antibody was developed using a bacterial fusion protein of MRP3 containing the segment of 830-949 AA.
Isotype IgG2b
Purification Tissue Culture Supernatant
Reactivity Human
Shipping Temperature Ambient RT
Storage temperature Store between 2°C and 8°C.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty