Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TrippGo™ Disposable Carts, TrippNT

Supplier: TRIPPNT
51706 51705 51708 51707 51709 51690 51692 51691 51687 51698 51700 51711 51689 51710 51688 51699 51702 51713 51701 51712 51704 51715 51714 51703
TrippNT's TrippGo Disposable Cart™ innovative design creates strength and function providing mobility and disposability

  • Available in 3 colors and 2 configuations
  • Disposable cart, smooth rolling casters, 2 locking
  • Built in tray on top
  • Durable Multi-layered cardboard construction
  • Comes with 3 drawers and bulk storage area or 5 drawers
  • Comes in white, brown, easily identifiable
  • Lightweight saves on shipping costs
  • Full cart capacity: 60 pounds
  • Ships flat, easy assembly
  • Eliminates need for costly decontamination or cleaning
  • Dispose of as required when your project is complete
  • Made in US

TrippGo disposable drawers fit TrippGo™ disposable carts and come in 2 sizes and colors; small, large, white or brown. Made from a versatile multi-layered cardboard. Lightweight. Keep fully stocked for excess storage and easy replenishment, or use as replacments. Weight capacity is 12 pounds per drawer. Easy assembly. Ships flat. Reuse or dispose as appropriate.

Made from a versatile multi-layered cardboard with 3 or 5 fully removable drawers. This lightweight easy-to-ship-flat cart has built-in tray on top, and smooth rolling casters. Use for hard-to-clean situations and then reuse or dispose of as appropriate. Perfect storage, mobility, lightweight access to supplies for field operations, hazardous environments and other temporary occasions. Easy assembly. Ships flat. Assembly takes only 5 minutes. Patent pending. Go anywhere with your TrippGo Disposable Cart!



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Shelf Capacity Compartment Dimensions Depth Drawer Dimensions Height Color No. of Drawers Weight Supplier No. VWR Catalog Number Unit Price Quantity
White, 3 Drawer 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" White 3 8.2 lbs. 51687 75783-964 Each Retrieving
Brown, 3 Drawer 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Brown 3 8.2 lbs. 51688 75783-966 Each Retrieving
Easily Identifiable, 3 Drawer 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Varied 3 8.2 lbs. 51689 75783-968 Each Retrieving
White, 5 Drawer 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" White 5 9.5 lbs. 51690 75783-974 Each Retrieving
Brown, 5 Drawer 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Brown 5 9.5 lbs. 51691 75783-976 Each Retrieving
Easily Identifiable, 5 Drawer 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Varied 5 9.5 lbs. 51692 75783-978 Each Retrieving
White, 3 Drawer, 3 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" White 3 8.2 lbs. 51698 75783-980 Pack of 1 Retrieving
Brown, 3 Drawer, 3 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Brown 3 8.2 lbs. 51699 75783-982 Pack of 1 Retrieving
Easily Identifiable, 3 Drawer, 3 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Varied 3 8.2 lbs. 51700 75783-984 Pack of 1 Retrieving
White, 5 Drawer, 3 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" White 5 9.5 lbs. 51701 75783-986 Pack of 1 Retrieving
Brown, 5 Drawer, 3 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Brown 5 9.5 lbs. 51702 75783-988 Pack of 1 Retrieving
Easily Identifiable, 5 Drawer, 3 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Varied 5 9.5 lbs. 51703 75783-990 Pack of 1 Retrieving
White, 3 Drawer, 10 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" White 3 8.2 lbs. 51704 75783-992 Pack of 1 Retrieving
Brown, 3 Drawer, 10 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Brown 3 8.2 lbs. 51705 75783-994 Pack of 1 Retrieving
Easily Identifiable, 3 Drawer, 10 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Varied 3 8.2 lbs. 51706 75783-996 Pack of 1 Retrieving
White, 5 Drawer, 10 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" White 5 9.5 lbs. 51707 75783-998 Pack of 1 Retrieving
Brown, 5 Drawer, 10 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Brown 5 9.5 lbs. 51708 75784-000 Pack of 1 Retrieving
Easily Identifiable, 5 Drawer, 10 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Varied 5 9.5 lbs. 51709 75784-002 Pack of 1 Retrieving
White, 3 Drawer, 50 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" White 3 8.2 lbs. 51710 75784-004 Pack of 1 Retrieving
Brown, 3 Drawer, 50 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Brown 3 8.2 lbs. 51711 75784-006 Pack of 1 Retrieving
Easily Identifiable, 3 Drawer, 50 Pack 12 lbs. per drawer, 24 lbs. bulk area Bulk Storage Area 12 x 141/2 x 153/16" 17" 111/2 x 3 x 147/8" 34" Varied 3 8.2 lbs. 51712 75784-008 Pack of 1 Retrieving
White, 5 Drawer, 50 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" White 5 9.5 lbs. 51713 75784-010 Pack of 1 Retrieving
Brown, 5 Drawer, 50 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Brown 5 9.5 lbs. 51714 75784-012 Pack of 1 Retrieving
Easily Identifiable, 5 Drawer, 50 Pack 12 lbs. per drawer 17" Small Drawers 111/2 x 3 x 147/8";
Large Drawers 111/2 x 61/2 x 147/8"
34" Varied 5 9.5 lbs. 51715 75784-014 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description For Use With Supplier No. VWR Catalog Number Unit Price Quantity
Small White Drawers, 3 Pack Carts 51687, 51688, 51689, 51690, 51691 and 51692 51716 75784-016 Pack of 1 Retrieving
Small Brown Drawers, 3 Pack Carts 51687, 51688, 51689, 51690, 51691 and 51692 51717 75784-018 Pack of 1 Retrieving
Large White Drawers, 2 Pack Carts 51687, 51688, 51689, 51690, 51691 and 51692 51718 75784-020 Pack of 1 Retrieving
Large Brown Drawers, 2 Pack Carts 51687, 51688, 51689, 51690, 51691 and 51692 51719 75784-022 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty