Specifications
- Size:4 x 25 mL
- Description:NEFA-HR(2) Color B
- Method:Enzymatic
- Storage temperature:2-10°C
- Linearity:0.01 - 4.00 mEq/L
- Cat. no.:CA97000-014
Specifications
About this item
The Wako HR series NEFA-HR(2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum.
See order tab for additional NEFA HR(2) reagents, linearity set, and calibrators.