Shake 'N' Stack™ Hybridization Ovens, Thermo Scientific
Supplier: Thermo Scientific
6243
CA11201-264EA 28456.62 CAD
CA11201-264
Shake 'N' Stack™ Hybridization Ovens, Thermo Scientific
Ovens Hybridisation Ovens
Conserve valuable space in your lab with these ovens, featuring a stackable, space-saving design with excellent uniformity ideal for molecular biology labs.
  • Accurate temperature control for improved experimental results
  • Excellent temperature uniformity for reproducible results with low backgrounds
  • Stackable format allows better utilization of laboratory space
  • Variable speed settings for protocol optimization
Multiple rotisserie fittings for flexible choice of consumable Interchangeable rotisserie/shaking platform for hybridization and washing procedures.
Delivery information: Includes10-bottle capacity rotisserie, adjustable feet, drip tray, manual.