Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CD40L [CD154] Recombinant Protein, Prosci

Supplier: Prosci
90-395
CD40 Ligand (CD40L), renamed TNFSF5 but now also known as CD154, TRAP and gp39, is a 34-39kDa type II transmembrane glycoprotein that belongs to the TNF superfamily. As with other TNF superfamily members, CD40L will exist as a trimer, both as a membrane bound and soluble form. Multiple mutations and alternate splice forms of CD40L exist, often associated with pathology and leading to truncated or nontrimerizable forms of CD40L. CD40L binds to both CD40 and to integrin alphaIIbbeta3 (CD41). In the cell membrane, it also associates with a unique splice variant of CD28 (CD28i) that may facilitate CD40L signal transduction. CD40L is expressed by monocytes, NK cells, mast cells, basophils, smooth muscle cells, endothelial cells, dendritic cells,activated and resting B cells, plus activated platelets and CD4+ T cells. CD40L ligation of CD40 on dendritic cells (DC) initiates DC maturation and differentiation. CD40L signaling into naive B cells promotes germinal center formation and isotope switching. CD40-CD40L seems to bridge innate and adaptive immune signals.

This recombinant proteins is for research use only.

Fusion-Tag: Fc Tag




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 μg 90-395 75794-216 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/peptide name CD40 Ligand
Protein synonyms Gp39, CD154, Tumor Necrosis Factor Ligand Superfamily Member 5,TNFSF5
Protein/peptide type Recombinant
Species Human
Source CHO cells
Sequence 113-261
Purity >98% (SDS-PAGE)
Formulation Lyophilized from 0.2um-filtered solution in PBS.
Storage conditions Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
Modification Fc Tag

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty