|
![]() |
ORDER
SPECIFICATIONS
Protein/peptide name | CD152 |
Protein synonyms | CTLA-4 |
Protein/peptide type | Recombinant |
Species | Mouse |
Source | NS1 cells |
Sequence | 38-160 |
Purity | >98% (SDS-PAGE) |
Formulation | Lyophilized from 0.2um-filtered solution in PBS. |
Biological activity | Binds both CD80 (B7-1) and CD86 (B7-2) with high affinity and inhibits CD28 signaling competitively. Kills the target cell completely. |
Storage conditions | Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C. |
Protein type | Recombinant |
Modification | Fc Tag |