Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CD200 non-lytic Recombinant Protein, Prosci

Supplier: Prosci
90-421
CD200, also known as OX-2, is a 45 kDa transmembrane immunoregulatory protein that belongs to the immunoglobulin superfamily. CD200 is widely but not ubiquitously expressed. Its receptor (CD200R) is restricted primarily to mast cells, basophils, macrophages, and dendritic cells, which suggests myeloid cell regulation as the major function of CD200. CD200 knockout mice are characterized by increased macrophage number and activation and are predisposed to autoimmune disorders. In T cells, CD200 functions as a costimulatory molecule independent of the CD28 pathway. Several viruses encode CD200 homologs which are expressed on infected cells during the lytic phase. Like CD200 itself, viral CD200 homologs also suppress myeloid cell activity, enabling increased viral propagation.

This recombinant proteins is for research use only.

Fusion-Tag: Fc Tag




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 90-421 75794-268 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/peptide name CD200 non-lytic
Protein synonyms OX-2 Membrane Glycoprotein, MOX1, MOX2
Protein/peptide type Recombinant
Species Human
Source CHO cells
Sequence 31-236
Purity >98% (SDS-PAGE)
Formulation Lyophilized from 0.2um-filtered solution in PBS.
Biological activity Shows the biological function of the CD200 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
Storage conditions Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
Protein type Recombinant
Modification Fc Tag
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty