0.33/0.74 mm Vu-Union Connector Replacement Inserts, Restek
Supplier: Restek
20419
RK20419PK 122.4 CAD
RK20419
0.33/0.74 mm Vu-Union Connector Replacement Inserts, Restek
Connectors Column Connectors
These replacements inserts are for use with Vu-Union connectors for GC-MS applications.
  • Fit columns with outside diameters of 0.33 or 0.74 mm
Vu-Union connector inserts are constructed from durable glass.