Universal Angled 'Y' Press-Tight® Connectors, Restek
Supplier: Restek
20403 20404-261 20469 20403-261 20487 20404
RK20403EA 128.51 CAD
RK20403 10852-950 RK20469 10852-972 10852-954 RK20404
Universal Angled 'Y' Press-Tight® Connectors, Restek
Connectors Column Connectors
Universal angled "Y" Press-Tight® connectors feature a split column flow to two detectors.
  • Inlet and outlet ends match column curvature to alleviate connection strain
  • Deactivated fused quartz helps eliminate activity for complete inertness along the GC flow path
  • Universal fit: use with 0.1–0.53 mm ID Restek columns (0.33–0.74 mm OD)
Optional Siltek® deactivation is ideal for organochlorine pesticides analysis.