ContactUsLinkComponent
Fluorometric & Colorimetric ADP Quantitation Kit, PromoKine
Fluorometric & Colorimetric ADP Quantitation Kit, PromoKine
Catalog #: 10180-428
Supplier:  Avantor
Fluorometric & Colorimetric ADP Quantitation Kit, PromoKine
Catalog #: 10180-428
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fluorometric & Colorimetric ADP Quantitation Kit
  • Size:
    100 assays
  • Cat. no.:
    10180-428
Specifications
About this item
PromoKine’s new Fluorometric & Colorimetric ADP Quantitation Kit provides a convenient means to measure the ADP level.
  • Simple, sensitive, stable and high-throughput adaptable
  • Can detect as low as 1 µM ADP in biological samples
  • ADP is converted to ATP and pyruvate
Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, the luciferase system is unstable and luminescence equipment is not generally available in most laboratories. PromoKine’s new Fluorometric & Colorimetric ADP Quantitation Kit provides a convenient colorimetric and fluorometric means to measure the ADP level. In this assay, ADP is converted to ATP and pyruvate. The generated pyruvate can be quantified colorimetrically (λmax = 570 nm) or fluorometrically (Ex/Em 535/587 nm). The assay is simple, sensitive, stable and high-throughput adaptable and can detect as low as 1 µM ADP in biological samples.
Customers Who Bought This Also Bought