Cytosol/Particulate Fractionation Kit, PromoKine
Supplier: PromoKine
PK-CA577-K267
10183-702EA 877.46 CAD
10183-702
Cytosol/Particulate Fractionation Kit, PromoKine
Cell Separation Media
The Cytosol/Particulate Separation Kit provides an easy and convenient system for a rapid separation of cytosol and particulate fractions.
  • Compatible with downstream applications
  • Assay cellular distributions of small molecules
  • Examine other signal transduction pathways
PromoKine's Cell Fractionation and Separation Kits contain unique formulations of buffers and reagents for convenient preparation of highly enriched cellular fractions (mitochondria, cytosol, nuclear) from mammalian cells. The Cytosol/Particulate Separation Kit provides an easy and convenient system for a rapid separation of cytosol and particulate fractions. The separated fractions are compatible with downstream applications, such as assaying cellular distributions of small molecules or proteins and examining other signal transduction pathways.