Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti- Mouse Monoclonal Antibody

Supplier: Genetex
GTX60410
Mouse Monoclonal Antibody [6F3] to E7

Type: Primary
Antigen: E7
Clonality: Monoclonal
Clone: 6F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: HPV




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl GTX60410 10168-182 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol E7
Antigen name E7
Reactivity HPV
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 6F3
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Gene ID 1489079
Immunogen The immunogen corresponding to E7 expressed in E. Coli.
Storage buffer Ascitic fluid containing 0.03% sodium azide.
Storage temperature 4°C; -20°C for long term storage
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty