Blade Carriers, Thermo Scientific
Supplier: Thermo Scientific
705040 705010 705040
CA84000-444EA 33205.01 CAD
CA84000-444 84000-442 84000-444
Blade Carriers, Thermo Scientific
Microtomes and Microtome Assemblies Blades and Handles
Insert a variety of Thermo Scientific™ blades in Thermo Scientific™ Blade Carriers.
  • Blade angle and blade adjustment levers are removable to help protect user settings
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
  • Compatible only with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
  • Standard Knife Carriers feature integrated knife guard with clearance angle adjustment
  • Complete with knife carrier base
  • Backlighting system adaptable
  • Laterally movable to accommodate conventional knives, tungsten carbide knives, and disposable blade inserts
  • Blade Holder Insert for low-profile blades to be inserted in the standard knife carriers N and SN (includes Allen key)
  • Knife Carrier Base with clamping device for use along with the upper parts of the knife carriers N, SN, NR, ER, C, SL