Order Entry
ContactUsLinkComponent
FiberFlo Hollow Fiber Cartridge Filter, Tek Products
FiberFlo Hollow Fiber Cartridge Filter, Tek Products
Catalog #: 89131-964
FiberFlo Hollow Fiber Cartridge Filter, Tek Products
Catalog #: 89131-964
Discontinued

Specifications

  • Maximum Flow Rate:
    2.5 GPM
  • Pore size:
    0.05 µm
  • Description:
    FiberFlo 50 Grade HF Cartridge Filter, 25.4 cm (10") length, 6.4 cm (2.9") diameter
  • Cat. no.:
    89131-964
  • Maximum Pressure Differential:
    30 psi (2 bar)

Specifications

About this item

The FiberFlo hollow fiber (HF) cartridge filter provides consistent and verifiable pyrogen removal.

  • Medical and pharmaceutical grade design
  • Wide choice of end connections
  • Higher flow rates at lower pressure drops
  • Extended life and repeatedly rewettable without wetting agents

With up to three times the surface area of comparable pleated cartridge filters, the HF filter provides greater capacity and longer life than pleated filters, resulting in lower filter replacement costs. It also offers a combination of higher surface area and membrane porosity.

High level endotoxin, bacteria, and spore removal makes this filter ideal for a range of water purification or process fluid applications.

Customers Who Bought This Also Bought