Rat Adipocyte Differentiation Media, Genlantis
Supplier: Genlantis
SR811D250
10497-738EA 232.79 CAD
10497-738
Rat Adipocyte Differentiation Media, Genlantis
Cell Separation Media
Characterized for bone mineralization and lipid accumulation following differentiation using adipocyte differentiation media.
  • Isolated from adult or fetal bone marrow
  • Cryopreserved at second passage
  • Lipid accumulation following differentiation using adipocyte differentiation media
Rat Marrow Stromal Cell (RMSC) are isolated from adult or fetal bone marrow. They are cryopreserved at second passage and can be cultured and propagated for 10 population doublings.
Rat Marrow Stromal Cell (RMSC) or also known as Mesenchymal stem cells (MSC), have the ability to self-renew and differentiate into cells of various connective tissue lineages, such as bone, cartilage, fat, and neurons. The potential of MSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue. Each lot of PrimaPure MSC's is tested for its ability to attach and spread on standard tissue culture grade plastic surfaces and proliferate in Rat Marrow Stromal Cell Growth Medium. Additionally, each lot is characterized for bone mineralization and lipid accumulation following differentiation using osteoblast and adipocyte differentiation media, repectively.