ContactUsLinkComponent
undefined | Additives and Supplements for Microbiology
Catalog #: CA10789-716
Catalog #: CA10789-716
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mayer's Albumin, HDx
  • Cat. no.:
    CA10789-716
  • Volume:
    15 mL
Specifications
About this item
Mayer's Albumin for better specimen to slide adhesion.
Used to adhere fecal specimens to glass slides in the Modified Iron Hematoxylin/Kinyoun or Trichrome staining process.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Customers Who Bought This Also Bought