Digital Timing System with Photogates

Supplier: VWR International
LBTM
470004-892EA 738.41 CAD
470004-892
Digital Timing System with Photogates
Timers Digital Timers
Microprocessor-controlled digital timer has eight built-in measurement modes.

  • Suitable for use with free fall apparatus
  • Displays LED 14 mm
  • Voltage in 110 V

This is a precision digital timer used to measure the time interval between any two events with high accuracy. The timer standard accuracy has been achieved using the 10 MHz Quartz Crystal Oscillator. It has four operating modes manual, free fall mode, timing gates mode and any other triggering start and stop. It has a built in power source for timing gates and the coil for g by free fall.

Delivery information: Supplied with two timing gates and manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR