Organochlorine Pesticide Mix C #1 (6 Components), Restek
Supplier: Restek
32296
75834-942EA 83.83 CAD
75834-942
Organochlorine Pesticide Mix C #1 (6 Components), Restek
Standards Organic Standards
Contains six components, those are chlorobenzilate, diallate, 1,2-dibromo-3-chloropropane, hexachlorobenzene, hexachlorocyclopentadiene and isodrin.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.