Acrolein, Restek
Supplier: Restek
Certificates
About this item
Ordering: This product has a limited shelf life. We recommend that you order only the ampoule quantity that meets your immediate needs.
Caution: NOTE: Store standards under frozen conditions (0 °C or colder). We recommend you store ampoules lying flat (horizontally) to prevent breakage. For best results, thaw the contents of the ampoule fully and shake well before opening and using the solution.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.