Would you like to log in again?
CHEMWARE® PTFE Stirrers, Saint Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069094 D1069093
These stirrers are ideal for precipitates and residues.
  • Non-stick, easy to clean
  • Low and high temperature resistant
Chisel tip on one end diagonal cut on other end permits applications in confined vessels
Unbreakable and heat resistant to 550 °F (288 °C), they will not crack or break under several conditions, such as thermal and mechanical shock.
Will not scratch glassware. Chemically inert PTFE offers resistance to adhesion of practically any material.
ORDER
Expand All / Collapse All
Length Supplier No. VWR Catalog Number Unit Price Quantity
15 cm (6") D1069093 53803-080 Each Retrieving
20 cm (8") D1069094 53803-116 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty