Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pouring Rings, Chemglass

Supplier: Chemglass
CLS-1482-17 CLS-1482-16 CLS-1482-15 CLS-1482-14 CLS-1482-13 CLS-1482-12 CLS-1482-11
These pour rings are made of high temperature ETFE (Ethylene Tetrafluoroethylene).

  • Steam autoclavable to 140 °C
  • Pouring rings are polypropylene
  • Max temperature is 180 °C



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Pouring Ring, GL-32, Blue, Polypropylene CLS-1482-11 30623-028 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-32, Clear, Polypropylene CLS-1482-12 30623-030 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-45, Blue, Polypropylene CLS-1482-13 30623-032 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-45, Green, Polypropylene CLS-1482-14 30623-034 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-45, Gray, Polypropylene CLS-1482-15 30623-036 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-45, Yellow, Polypropylene CLS-1482-16 30623-038 Pack of 10 Retrieving
Small Business Enterprise
Pouring Ring, GL-45, Clear, Polypropylene CLS-1482-17 30623-040 Pack of 10 Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping